APO455825666 Apollo Scientific Ltd

4-Bromophenylhydrazine

  • MDL number MFCD01935685
  • Cas Number : 589-21-9
  • Molecular Weight : 187.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 102-104
Iupac Name 1-(4-bromophenyl)hydrazine
Inchi Code 1S/C6H7BrN2/c7-5-1-3-6(9-8)4-2-5/h1-4,9H,8H2
Inchi Key NRESDXFFSNBDGP-UHFFFAOYSA-N

Description

Synonym 1-Bromo-4-hydrazinobenzene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 102-104
Product Link https://store.apolloscientific.co.uk/product/4-bromophenylhydrazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromophenylhydrazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart