APO455825671 Apollo Scientific Ltd

(4-Bromophenyl)(isocyano)methyl 4-methylphenyl sulphone

  • MDL number MFCD04114782
  • Cas Number : 655254-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-1-[isocyano(toluene-4-sulphonyl)]methylbenzene
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39360

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39360
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart