APO455825680 Apollo Scientific Ltd

5-(2,4-Dichlorophenyl)-4H-pyrazole-3-carboxylic acid

  • MDL number MFCD06245447
  • Cas Number : 904815-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Carboxy-5-(2,4-dichlorophenyl)-4H-pyrazole
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39620

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39620
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart