APO455825681 Apollo Scientific Ltd

3,4-Dihydro-1H-1,4-benzodiazepine-2,5-dione

  • MDL number MFCD00111349
  • Cas Number : 5118-94-5
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 327-328
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/34-dihydro-1h-14-benzodiazepine-25-dione

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/34-dihydro-1h-14-benzodiazepine-25-dione
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart