APO455825684 Apollo Scientific Ltd

1-(2-Nitrophenyl)imidazolidin-2-one

  • MDL number MFCD02671217
  • Cas Number : 500890-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(2-Nitrophenyl)tetrahydro-2H-imidazol-2-one
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39684

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39684
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart