APO455825689 Apollo Scientific Ltd

2-(3,5-Dimethyl-1H-pyrazol-1-yl)benzoic acid

  • MDL number MFCD06245567
  • Cas Number : 27363-77-5
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-35-dimethyl-1h-pyrazol-1-ylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-35-dimethyl-1h-pyrazol-1-ylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart