APO455825698 Apollo Scientific Ltd

Methyl 5-phenyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD00781977
  • Cas Number : 56426-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-182
Synonym 3-(Methoxycarbonyl)-5-phenyl-1H-pyrazole, [3-(Methoxycarbonyl)-1H-pyrazol-5-yl]benzene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/methyl-5-phenyl-1h-pyrazole-3-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-phenyl-1h-pyrazole-3-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart