APO455825700 Apollo Scientific Ltd

2-(2-Nitrophenyl)-1H-benzimidazole-5-carboxylic acid

  • MDL number MFCD01159447
  • Cas Number : 190121-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Carboxy-2-(2-nitrophenyl)-1H-benzimidazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=39799

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=39799
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart