APO455825702 Apollo Scientific Ltd

4-Bromo-1-methyl-1H-pyrazole-3-carboxaldehyde

  • MDL number MFCD00103235
  • Cas Number : 287917-96-8
  • Molecular Weight : 189.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-76
Synonym 4-Bromo-3-formyl-1-methyl-1H-pyrazole
Purity 95
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-1-methyl-1h-pyrazole-3-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-1-methyl-1h-pyrazole-3-carboxaldehyde
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart