APO455825706 Apollo Scientific Ltd

Imidazo[1,2-b]pyridazine

  • MDL number MFCD07782103
  • Cas Number : 766-55-2
  • Molecular Weight : 119.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-54
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/imidazo12-bpyridazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/imidazo12-bpyridazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart