APO455825708 Apollo Scientific Ltd

2-Chloro-6-phenylnicotinonitrile

  • MDL number MFCD01316320
  • Cas Number : 43083-14-3
  • Molecular Weight : 214.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-6-phenylpyridine-3-carbonitrile, 2-Chloro-3-cyano-6-phenylpyridine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-phenylnicotinonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-phenylnicotinonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart