APO455825709 Apollo Scientific Ltd

2-(1H-Pyrrol-1-yl)benzylamine

  • MDL number MFCD03763920
  • Cas Number : 39243-88-4
  • Molecular Weight : 172.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [2-(1H-Pyrrol-1-yl)phenyl]methylamine, 1-[2-(Aminomethyl)phenyl]-1H-pyrrole
Purity 95
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 80-82/0.15mm
Product Link https://store.apolloscientific.co.uk/product/2-1h-pyrrol-1-ylbenzylamine

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-1h-pyrrol-1-ylbenzylamine
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart