APO455825714 Apollo Scientific Ltd

6-Chloro-2-(piperazin-1-yl)-1,3-benzothiazole

  • MDL number MFCD01571353
  • Cas Number : 153025-29-7
  • Molecular Weight : 253.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(6-Chloro-1,3-benzothiazol-2-yl)piperazine
Purity 95
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-chloro-2-piperazin-1-yl-13-benzothiazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-chloro-2-piperazin-1-yl-13-benzothiazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart