APO455825723 Apollo Scientific Ltd

5-Oxo-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid

  • MDL number MFCD01763813
  • Cas Number : 3470-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215-222(dec.)
Synonym 6-Carboxy-1-oxo-1,2,3,4-tetrahydronaphthalene, 5-Oxo-5,6,7,8-tetrahydro-2-naphthoic acid
Purity 96
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28367

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28367
pictograms GHS05;GHS07
hstatements H302;H312;H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart