APO455825724 Apollo Scientific Ltd

N-(4-Bromobut-1-yl)-4-nitrophthalimide

  • MDL number MFCD01313849
  • Cas Number : 125207-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-104
Synonym 2-(4-Bromobut-1-yl)-5-nitro-1H-isoindole-1,3(2H)-dione, 2-(4-Bromobut-1-yl)-5-nitroisoindolin-1,3-dione
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13181

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13181
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart