APO455825727 Apollo Scientific Ltd

1-(But-3-en-1-yl)-3-chlorobenzene

  • MDL number MFCD01319546
  • Cas Number : 91426-46-9
  • Molecular Weight : 166.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(3-Chlorophenyl)but-1-ene, 1-Chloro-3-homoallylbenzene
Physical Form Liquid-Oil
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-but-3-en-1-yl-3-chlorobenzene

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-but-3-en-1-yl-3-chlorobenzene
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart