APO455825728 Apollo Scientific Ltd

1-[(Pyridin-4-yl)methyl]piperidine-4-carboxylic acid

  • MDL number MFCD03645526
  • Cas Number : 774531-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 222-224
Synonym 4-[(4-Carboxypiperidin-1-yl)methyl]pyridine, 4-Carboxy-1-[(pyridin-4-yl)methyl]piperidine
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/1-pyridin-4-ylmethylpiperidine-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-pyridin-4-ylmethylpiperidine-4-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart