APO455825729 Apollo Scientific Ltd

2-Acetyl-4-nitroindane-1,3-dione

  • MDL number MFCD00728684
  • Cas Number : 25125-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-148
Synonym 2-Acetyl-4-nitro-1H-indene-1,3(2H)-dione, 2-Acetyl-2,3-dihydro-4-nitro-1,3-dioxo-1H-indene
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12434

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12434
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart