APO455825731 Apollo Scientific Ltd

Ethyl 1,2,4-oxadiazole-3-carboxylate

  • MDL number MFCD06245497
  • Cas Number : 39512-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41-43
Synonym 3-(Ethoxycarbonyl)-1,2,4-oxadiazole
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 168-171/1mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28370

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28370
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart