APO455825741 Apollo Scientific Ltd

Methyl 3-amino-2-hydroxybenzoate

  • MDL number MFCD09842614
  • Cas Number : 35748-34-6
  • Molecular Weight : 167.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 86-89
Iupac Name methyl 3-amino-2-hydroxybenzoate
Inchi Code 1S/C8H9NO3/c1-12-8(11)5-3-2-4-6(9)7(5)10/h2-4,10H,9H2,1H3
Inchi Key OMWQHVRUXLRZRC-UHFFFAOYSA-N

Description

Synonym 2-Hydroxy-3-(methoxycarbonyl)aniline, 2-Amino-6-(methoxycarbonyl)phenol
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 86-89
Product Link https://store.apolloscientific.co.uk/product/methyl-3-amino-2-hydroxybenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-amino-2-hydroxybenzoate
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart