APO455825742 Apollo Scientific Ltd

5-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD09836168
  • Cas Number : 1000017-93-5
  • Molecular Weight : 196.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Carboxy-5-chloroimidazo[1,2-a]pyridine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloroimidazo12-apyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloroimidazo12-apyridine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart