APO455825743 Apollo Scientific Ltd

4,5-Dichloro-2-nitrophenol

  • MDL number MFCD09836170
  • Cas Number : 39224-65-2
  • Molecular Weight : 208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-70
Synonym 4,5-Dichloro-2-hydroxynitrobenzene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/45-dichloro-2-nitrophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/45-dichloro-2-nitrophenol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart