APO455825748 Apollo Scientific Ltd

3-Acetyl-2-methylimidazo[1,2-a]pyridine

  • MDL number MFCD00665892
  • Cas Number : 29096-60-4
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112
Synonym 1-(2-Methylimidazo[1,2-a]pyridin-3-yl)ethan-1-one
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-acetyl-2-methylimidazo12-apyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-acetyl-2-methylimidazo12-apyridine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart