APO455825749 Apollo Scientific Ltd

3-Amino-4-hydroxybenzonitrile

  • MDL number MFCD00234273
  • Cas Number : 14543-43-2
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-153
Synonym 5-Cyano-2-hydroxyaniline, 2-Amino-4-cyanophenol
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-4-hydroxybenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-4-hydroxybenzonitrile
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart