APO455825753 Apollo Scientific Ltd

3-Amino-5-bromo-1-methyl-1H-indazole

  • MDL number MFCD09864804
  • Cas Number : 1000018-06-3
  • Molecular Weight : 226.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-135
Synonym 5-Bromo-1-methyl-1H-indazol-3-amine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-5-bromo-1-methyl-1h-indazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-5-bromo-1-methyl-1h-indazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart