APO455825755 Apollo Scientific Ltd

4-Bromo-1-methyl-1H-pyrazole-3-amidoxime

  • MDL number MFCD00831610
  • Cas Number : 287922-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-184
Synonym 4-Bromo-N'-hydroxy-1-methyl-1H-pyrazole-3-carboximidamide, 4-Bromo-N'-hydroxy-1-methyl-1H-pyrazole-3-carboxamidine
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13353

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13353
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart