APO455825758 Apollo Scientific Ltd

5-(Chlorosulphonyl)-2-methylbenzoic acid

  • MDL number MFCD03989642
  • Cas Number : 89001-57-0
  • Molecular Weight : 234.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-154
Synonym 5-(Chlorosulphonyl)-o-toluic acid, 3-Carbonyl-4-methylbenzenesulphonyl chloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chlorosulphonyl-2-methylbenzoic-acid

Safety Information

signalword Danger
pcodes P280;P309+P311
MSDS https://store.apolloscientific.co.uk/product/5-chlorosulphonyl-2-methylbenzoic-acid
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart