APO455825764 Apollo Scientific Ltd

4-(Methylsulphonyl)phenylacetonitrile

  • MDL number MFCD00079775
  • Cas Number : 25025-07-4
  • Molecular Weight : 195.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122-124
Synonym 4-(Methylsulphonyl)benzyl cyanide, 4-(Cyanomethyl)phenyl methyl sulphone
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-methylsulphonylphenylacetonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylsulphonylphenylacetonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart