APO455825766 Apollo Scientific Ltd

2-[4-(tert-Butoxycarbonyl)piperazin-1-yl]benzoic acid

  • MDL number MFCD04115068
  • Cas Number : 444582-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 162-165
Synonym 4-(2-Carboxyphenyl)piperazine, N1-BOC protected, tert-Butyl 4-(2-carboxyphenyl)piperazine-1-carboxylate
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-4-tert-butoxycarbonylpiperazin-1-ylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-4-tert-butoxycarbonylpiperazin-1-ylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart