APO455825784 Apollo Scientific Ltd

2-Chloro-4-(methylsulphonyl)benzoic acid

  • MDL number MFCD00216496
  • Cas Number : 53250-83-2
  • Molecular Weight : 234.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191-196
Synonym 4-Carboxy-3-chlorophenyl methyl sulphone
Purity 97
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-methylsulphonylbenzoic-acid

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-methylsulphonylbenzoic-acid
pictograms GHS05;GHS07
hstatements H314;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart