APO455825792 Apollo Scientific Ltd

3-(4-Chlorophenyl)propanol

  • MDL number MFCD00831102
  • Cas Number : 6282-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 98-102/0.1mm
Product Link https://store.apolloscientific.co.uk/product/3-4-chlorophenylpropanol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-4-chlorophenylpropanol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart