APO455825802 Apollo Scientific Ltd

Cyclobutane-1,2-dicarboxylic acid anhydride

  • MDL number MFCD00799396
  • Cas Number : 4462-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-76
Synonym 3-Oxabicyclo[3.2.0]heptane-2,4-dione, 2,4-Dioxo-3-oxabicyclo[3.2.0]heptane
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/cyclobutane-12-dicarboxylic-acid-anhydride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/cyclobutane-12-dicarboxylic-acid-anhydride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart