APO455825811 Apollo Scientific Ltd

1,3-Diazaspiro[4.5]decane-2,4-dione

  • MDL number MFCD00022404
  • Cas Number : 702-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218-220
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/13-diazaspiro45decane-24-dione

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/13-diazaspiro45decane-24-dione
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart