APO455825816 Apollo Scientific Ltd

3,5-Dichlorobenzhydrazide

  • MDL number MFCD00833411
  • Cas Number : 62899-78-9
  • Molecular Weight : 205.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 205-207
Synonym 3,5-Dichlorobenzohydrazide
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-dichlorobenzhydrazide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-dichlorobenzhydrazide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart