APO455825820 Apollo Scientific Ltd

2,7-Dichloro-9H-fluorene

  • MDL number MFCD00032840
  • Cas Number : 7012-16-0
  • Molecular Weight : 235.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-128
Synonym 4,4'-Dichloro-2,2'-methylenebiphenyl
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/27-dichloro-9h-fluorene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/27-dichloro-9h-fluorene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart