APO455825828 Apollo Scientific Ltd

Diethyl 2-(2,4-dichlorophenyl)malonate

  • MDL number MFCD00831103
  • Cas Number : 111544-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Diethyl (2,4-dichlorophenyl)propanedioate
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 136-139/0.05mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14288

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14288
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart