APO455825829 Apollo Scientific Ltd

2,3-Dihydrobenzo[b]furan-5-amidoxime

  • MDL number MFCD04035721
  • Cas Number : 306936-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215-217
Synonym 2,3-Dihydro-N'-hydroxy-1-benzofuran-5-carboximidamide, 2,3-Dihydro-N'-hydroxybenzo[b]furan-5-carboxamidine
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14295

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14295
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart