APO455825830 Apollo Scientific Ltd

3-(4,5-Dihydro-4,4-dimethyl-1,3-oxazol-2-yl)pyridine

  • MDL number MFCD01313220
  • Cas Number : 68981-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4,5-Dihydro-4,4-dimethyl-2-(pyridin-3-yl)-1,3-oxazole
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14308

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14308
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart