APO455825834 Apollo Scientific Ltd

4-(Dimethylamino)phenyl isothiocyanate

  • MDL number MFCD00041093
  • Cas Number : 2131-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-70
Synonym N,N-Dimethyl-4-isothiocyanatoaniline
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 152/6mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14424

Safety Information

signalword Danger
pcodes P280;P260;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14424
pictograms GHS05;GHS06
hstatements H301;H311;H314;H330
Availability details for
Quantity
  • Availability
  • Add to Cart