APO455825843 Apollo Scientific Ltd

Ethyl 2-chloro-6-methylpyrimidine-4-carboxylate

  • MDL number MFCD00662808
  • Cas Number : 265328-14-1
  • Molecular Weight : 200.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-55
Synonym 2-Chloro-4-(ethoxycarbonyl)-6-methylpyrimidine, 2-Chloro-4-(ethoxycarbonyl)-6-methyl-1,3-diazine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-chloro-6-methylpyrimidine-4-carboxylate

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-chloro-6-methylpyrimidine-4-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart