APO455825845 Apollo Scientific Ltd

Ethyl 3,5-dichloro-4-hydroxybenzoate

  • MDL number MFCD00016420
  • Cas Number : 17302-82-8
  • Molecular Weight : 235.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 110-112
Iupac Name ethyl 3,5-dichloro-4-hydroxybenzoate
Inchi Code 1S/C9H8Cl2O3/c1-2-14-9(13)5-3-6(10)8(12)7(11)4-5/h3-4,12H,2H2,1H3
Inchi Key WMKNGSJJEMFQOT-UHFFFAOYSA-N

Description

Synonym 2,6-Dichloro-4-(ethoxycarbonyl)phenol
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 110-112
Product Link https://store.apolloscientific.co.uk/product/ethyl-35-dichloro-4-hydroxybenzoate

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/ethyl-35-dichloro-4-hydroxybenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart