APO455825850 Apollo Scientific Ltd

2-Ethyl-6-methylphenol

  • MDL number MFCD00962292
  • Cas Number : 1687-64-5
  • Molecular Weight : 136.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Ethyl-2-hydroxytoluene, 6-Ethyl-o-cresol
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 96-98/15mm
Product Link https://store.apolloscientific.co.uk/product/2-ethyl-6-methylphenol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-ethyl-6-methylphenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart