APO455825851 Apollo Scientific Ltd

Ethyl 2-phenylpropanoate

  • MDL number MFCD00795471
  • Cas Number : 2510-99-8
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl hydratropate
Inchi Code 1S/C11H14O2/c1-3-13-11(12)9(2)10-7-5-4-6-8-10/h4-9H,3H2,1-2H3
Inchi Key UTUVIKZNQWNGIM-UHFFFAOYSA-N

Description

Synonym Ethyl 2-phenylpropionate, Ethyl hydratropate
Flash Point 122/20mm
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-2-phenylpropanoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-2-phenylpropanoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart