APO455825852 Apollo Scientific Ltd

2-(1,3-Benzodioxol-5-yl)-2-chloroacetamide

  • MDL number MFCD08445595
  • Cas Number : 873380-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-(2-Amino-1-chloro-2-oxoethyl)-1,3-benzodioxole, 2-Chloro-2-[3,4-(methylenedioxy)phenyl]acetamide
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28989

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28989
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart