APO455825866 Apollo Scientific Ltd

4-Iodo-2-nitrotoluene

  • MDL number MFCD00051090
  • Cas Number : 41252-97-5
  • Molecular Weight : 263.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59-61
Synonym 4-Iodo-1-methyl-2-nitrobenzene, 5-Iodo-2-methylnitrobenzene
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-iodo-2-nitrotoluene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-iodo-2-nitrotoluene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart