APO455825870 Apollo Scientific Ltd

4-Iodophenyl isothiocyanate

  • MDL number MFCD00041091
  • Cas Number : 2059-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-78
Synonym 1-Iodo-4-isothiocyanatobenzene
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14967

Safety Information

signalword Warning
pcodes P280;P260;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14967
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart