APO455825874 Apollo Scientific Ltd

1-Isopropyl-1H-benzotriazole-5-carboxylic acid

  • MDL number MFCD01570664
  • Cas Number : 306935-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229-230
Synonym 1-(Prop-2-yl)-1H-benzotriazole-5-carboxylic acid, 5-Carboxy-1-isopropyl-1H-benzotriazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14996

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14996
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart