APO455825875 Apollo Scientific Ltd

2-Isopropyl-6-methylphenyl isothiocyanate

  • MDL number MFCD00060370
  • Cas Number : 306935-86-4
  • Molecular Weight : 191.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Isothiocyanato-1-methyl-3-(prop-2-yl)benzene, 3-Isopropyl-2-isothiocyanatotoluene
Density 1.02
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 92/3mm
Product Link https://store.apolloscientific.co.uk/product/2-isopropyl-6-methylphenyl-isothiocyanate

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/2-isopropyl-6-methylphenyl-isothiocyanate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart