APO455825887 Apollo Scientific Ltd

Methyl 3,4-dichlorophenylacetate

  • MDL number MFCD00800680
  • Cas Number : 6725-44-6
  • Molecular Weight : 219.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 25-27
Synonym 3,4-Dichlorophenylacetic acid methyl ester
Density 1.33
Physical Form Solid-Low Melt
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-34-dichlorophenylacetate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-34-dichlorophenylacetate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart