APO455825891 Apollo Scientific Ltd

2-Methyl-5-nitrobenzenesulphonamide

  • MDL number MFCD00115503
  • Cas Number : 6269-91-6
  • Molecular Weight : 216.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197-199
Synonym 4-Nitro-2-sulphamoyltoluene, 3-(Aminosulphonyl)-4-methylnitrobenzene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-methyl-5-nitrobenzenesulphonamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-methyl-5-nitrobenzenesulphonamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart